3-Fluor-4-methylthiobenzamid

Lieferant: Apollo Scientific
Gefahr

PC32663-1G
APOSPC32663-1GEA 149 EUR
APOSPC32663-1G
3-Fluor-4-methylthiobenzamid
3-Fluor-4-methylthiobenzamid
Formel: C₈H₈FNS
Molecular Weight: 169,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00275549
CAS-Nummer: 175277-87-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR