N-(4-Chlorophenyl)-2,6-dichloro-5-fluoronicotinamide
Lieferant: Apollo Scientific
|
Achtung
|
PC32683-250MG
APOSPC32683-250MGEA
33.5
EUR
APOSPC32683-250MG
N-(4-Chlorophenyl)-2,6-dichloro-5-fluoronicotinamide
N-(4-Chlorophenyl)-2,6-dichloro-5-fluoronicotinamide
Formel:
C₁₂H₆Cl₃FN₂O Schmelzpunkt: 159…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01765453 CAS-Nummer: 680217-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...