4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,6-Dichloro-5-fluoropyridin-3-yl)(morpholino)methanone

PC32691-250MG
APOSPC32691-250MGEA 79.9 EUR
APOSPC32691-250MG
4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine
4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine
Formel: C₁₀H₉Cl₂FN₂O₂
Molecular Weight: 279,095 g/mol
Schmelzpunkt: 132…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01765490
CAS-Nummer: 680217-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR