2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide

Lieferant: Apollo Scientific
Achtung

PC32692-100MG
APOSPC32692-100MGEA 29 EUR
APOSPC32692-100MG
2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide
2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide
Formel: C₁₁H₆Cl₂FN₃O
Schmelzpunkt: 133…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01934909
CAS-Nummer: 680217-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR