2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide

Lieferant: Apollo Scientific
Achtung

PC32711-1G PC32711-5G
APOSPC32711-1GEA 279 EUR
APOSPC32711-1G APOSPC32711-5G
2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide
2-Bromo-2,2-difluoro-N-(2-hydroxyphenyl)acetamide
Formel: C₈H₆BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11035865
CAS-Nummer: 239136-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR