1-Methyl-3-(trifluoroacetyl)-1H-indole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-one

PC32714-1G PC32714-5G
APOSPC32714-1GEA 69.6 EUR
APOSPC32714-1G APOSPC32714-5G
1-Methyl-3-(trifluoroacetyl)-1H-indole
1-Methyl-3-(trifluoroacetyl)-1H-indole
Formel: C₁₁H₈F₃NO
Molecular Weight: 227.183 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02571216
CAS-Nummer: 318-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR