2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Bis(trifluoroacetyl)-N,N-dimethylnaphth-1ylamine
PC32717-1G
PC32717-5G
APOSPC32717-1GEA
91.5
EUR
APOSPC32717-1G
APOSPC32717-5G
2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
2,4-Bis(trifluoroacetyl)-1-(N,N-dimethylamino)naphthalene
Formel:
C₁₆H₁₁F₆NO₂ Molecular Weight: 363.254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11035868 CAS-Nummer: 115975-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...