Ethyl difluoro(pyridin-2-yl)acetate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,2-difluoro-2-(pyridin-2-yl)acetate

PC32718-1G
APOSPC32718-1GEA 119 EUR
APOSPC32718-1G
Ethyl difluoro(pyridin-2-yl)acetate 95%
Ethyl difluoro(pyridin-2-yl)acetate
Formel: C₉H₉F₂NO₂
Molecular Weight: 201,17 g/mol
Siedepunkt: 79,5 °C/0.5mmHg
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06858718
CAS-Nummer: 267876-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR