4-{[1,3-Dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]oxy}benzaldehyde

Lieferant: Apollo Scientific

PC3272-1G
APOSPC3272-1GEA 277 EUR
APOSPC3272-1G
4-{[1,3-Dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]oxy}benzaldehyde
4-{[1,3-Dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]oxy}benzaldehyde
Formel: C₁₆H₁₂F₃N₃O₂
Molecular Weight: 335,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998164
CAS-Nummer: 1031928-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR