3-Fluoro-4-iodobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC32738-1G PC32738-250MG PC32738-5G
APOSPC32738-1GEA 0 EUR
APOSPC32738-1G APOSPC32738-250MG APOSPC32738-5G
3-Fluoro-4-iodobenzyl bromide
3-Fluoro-4-iodobenzyl bromide
Formel: C₇H₅BrFI
Lagertemperatur: Raumtemperatur
MDL: MFCD18396959
CAS-Nummer: 1022931-83-4
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR