4-Fluoro-2-formylphenylboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-2-formylbenzeneboronic acid, (4-Fluoro-2-formylphenyl)boronic acid

PC32757-1G PC32757-25G PC32757-5G
APOSPC32757-1GEA 21.9 EUR
APOSPC32757-1G APOSPC32757-25G APOSPC32757-5G
4-Fluoro-2-formylphenylboronic acid
4-Fluoro-2-formylphenylboronic acid
Formel: C₇H₆BFO₃
Schmelzpunkt: 123…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10697426
CAS-Nummer: 825644-26-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR