7-(Trifluoromethyl)chroman-4-one 97%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydro-7-(trifluoromethyl)-4H-chromen-4-one

PC3277-250MG
APOSPC3277-250MGEA 94.4 EUR
APOSPC3277-250MG
7-(Trifluoromethyl)chroman-4-one 97%
7-(Trifluoromethyl)chroman-4-one
Formel: C₁₀H₇F₃O₂
Molecular Weight: 216,157 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09998165
CAS-Nummer: 111141-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR