1-Bromo-2-chloro-3,5-difluorobenzene 98%

Lieferant: Apollo Scientific
Achtung

PC32780-10G PC32780-1G PC32780-5G
APOSPC32780-10GEA 104 EUR
APOSPC32780-10G APOSPC32780-1G APOSPC32780-5G
1-Bromo-2-chloro-3,5-difluorobenzene 98%
1-Bromo-2-chloro-3,5-difluorobenzene
Formel: C₆H₂BrClF₂
Molecular Weight: 227.434 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00135441
CAS-Nummer: 187929-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR