7-Bromo-5-fluorobenzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC32792-1G PC32792-250MG
APOSPC32792-1GEA 208 EUR
APOSPC32792-1G APOSPC32792-250MG
7-Bromo-5-fluorobenzofuran-2-carboxylic acid
7-Bromo-5-fluorobenzofuran-2-carboxylic acid
Formel: C₉H₄BrFO₃
Molecular Weight: 259.029 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13187183
CAS-Nummer: 550998-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR