4-Fluoro-2-methyl-5-nitroanisole

Lieferant: Apollo Scientific
Achtung

PC32802-1G PC32802-250MG PC32802-5G
APOSPC32802-1GEA 136 EUR
APOSPC32802-1G APOSPC32802-250MG APOSPC32802-5G
4-Fluoro-2-methyl-5-nitroanisole
4-Fluoro-2-methyl-5-nitroanisole
Formel: C₈H₈FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27937156
CAS-Nummer: 134882-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR