4'-Bromo-2',6'-difluoroacetophenone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-2,6-difluorophenyl)ethanone, 1-(4-Bromo-2,6-difluorophenyl)ethan-1-one

PC32806-250MG PC32806-1G PC32806-5G
APOSPC32806-250MGEA 21.9 EUR
APOSPC32806-250MG APOSPC32806-1G APOSPC32806-5G
4'-Bromo-2',6'-difluoroacetophenone 97%
4'-Bromo-2',6'-difluoroacetophenone
Formel: C₈H₅BrF₂O
Molecular Weight: 235.026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22570410
CAS-Nummer: 746630-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR