4,4-Difluorocyclohexanecarbonyl chloride
Lieferant: Apollo Scientific
Synonyme:
4,4-Difluorocyclohexane-1-carbonyl chloride
PC32810-1G
APOSPC32810-1GEA
122
EUR
APOSPC32810-1G
4,4-Difluorocyclohexanecarbonyl chloride
4,4-Difluorocyclohexanecarbonyl chloride
Formel:
C₇H₉ClF₂O Molecular Weight: 182,6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD13173983 CAS-Nummer: 376348-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...