3',5'-Difluor-4'-methoxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3,5-Difluoro-4-methoxyphenyl)ethanone

PC3283-1G
APOSPC3283-1GEA 62.5 EUR
APOSPC3283-1G
3',5'-Difluor-4'-methoxyacetophenon 98%
3',5'-Difluor-4'-methoxyacetophenon
Formel: C₉H₈F₂O₂
Molecular Weight: 186,15 g/mol
Schmelzpunkt: 27…31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236243
CAS-Nummer: 170570-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR