3-Iodo-4-(trifluoromethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC32839-1G PC32839-250MG PC32839-5G
APOSPC32839-1GEA 0 EUR
APOSPC32839-1G APOSPC32839-250MG APOSPC32839-5G
3-Iodo-4-(trifluoromethoxy)aniline
3-Iodo-4-(trifluoromethoxy)aniline
Formel: C₇H₅F₃INO
Lagertemperatur: Raumtemperatur
MDL: MFCD20731076
CAS-Nummer: 1365969-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR