2-(Chloromethyl)-5-(4-fluorophenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

PC32870-1G
APOSPC32870-1GEA 256 EUR
APOSPC32870-1G
2-(Chloromethyl)-5-(4-fluorophenyl)-1,3,4-oxadiazole
2-(Chloromethyl)-5-(4-fluorophenyl)-1,3,4-oxadiazole
Formel: C₉H₆ClFN₂O
Molecular Weight: 212,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05263166
CAS-Nummer: 350672-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR