N'-Hydroxy-2-[4-(trifluoromethyl)phenyl]ethanimidamide

Lieferant: Apollo Scientific
Achtung

PC32874-1G PC32874-250MG PC32874-50MG PC32874-500MG
APOSPC32874-1GEA 510 EUR
APOSPC32874-1G APOSPC32874-250MG APOSPC32874-50MG APOSPC32874-500MG
N'-Hydroxy-2-[4-(trifluoromethyl)phenyl]ethanimidamide
N'-Hydroxy-2-[4-(trifluoromethyl)phenyl]ethanimidamide
Formel: C₉H₉F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16077266
CAS-Nummer: 1312766-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR