2,4-Difluorophenyl-2-chloroacetate

Lieferant: Apollo Scientific

PC32875-1G
APOSPC32875-1GEA 386 EUR
APOSPC32875-1G
2,4-Difluorophenyl-2-chloroacetate
2,4-Difluorophenyl-2-chloroacetate
Formel: C₈H₅ClF₂O₂
Molecular Weight: 206,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001124
CAS-Nummer: 1312137-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR