6-(Trifluoromethyl)pyridine-2-carboxamide
Lieferant: Apollo Scientific
Synonyme:
6-(Trifluoromethyl)picolinamide
PC32880-1G
APOSPC32880-1GEA
78.3
EUR
APOSPC32880-1G
6-(Trifluoromethyl)pyridine-2-carboxamide
6-(Trifluoromethyl)pyridine-2-carboxamide
Formel:
C₇H₅F₃N₂O Molecular Weight: 190,123 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02180820 CAS-Nummer: 22245-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...