8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC32883-1G
APOSPC32883-1GEA
261
EUR
APOSPC32883-1G
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel:
C₇H₃ClF₃N₃ Molecular Weight: 221,567 g/mol Schmelzpunkt: 191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315052 CAS-Nummer: 478066-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...