4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine

Lieferant: Apollo Scientific

PC32884-1G PC32884-250MG PC32884-5G
APOSPC32884-1GEA 0 EUR
APOSPC32884-1G APOSPC32884-250MG APOSPC32884-5G
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
Formel: C₈H₅F₃N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00160038
CAS-Nummer: 60388-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR