Ethyl-2-(trifluoromethyl)phenylacetate 97%

Lieferant: Apollo Scientific
Achtung

PC3288D-1G
APOSPC3288D-1GEA 52.2 EUR
APOSPC3288D-1G
Ethyl-2-(trifluoromethyl)phenylacetate 97%
Ethyl-2-(trifluoromethyl)phenylacetate
Formel: C₁₁H₁₁F₃O₂
Molecular Weight: 232,2 g/mol
Flammpunkt: 66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236677
CAS-Nummer: 81580-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR