4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one

Lieferant: Apollo Scientific
Achtung

PC32899-1G PC32899-250MG
APOSPC32899-1GEA 84.1 EUR
APOSPC32899-1G APOSPC32899-250MG
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Formel: C₈H₄F₃N₃O
Molecular Weight: 215.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31742301
CAS-Nummer: 2379918-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR