4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Lieferant: Apollo Scientific
|
Achtung
|
PC32899-1G
PC32899-250MG
APOSPC32899-1GEA
84.1
EUR
APOSPC32899-1G
APOSPC32899-250MG
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3,4,5-Trifluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Formel:
C₈H₄F₃N₃O Molecular Weight: 215.132 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31742301 CAS-Nummer: 2379918-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...