5-Chloro-2-(2-fluoroethoxy)benzoic acid 99%

Lieferant: Apollo Scientific
Achtung

PC32901-100MG
APOSPC32901-100MGEA 69.6 EUR
APOSPC32901-100MG
5-Chloro-2-(2-fluoroethoxy)benzoic acid 99%
5-Chloro-2-(2-fluoroethoxy)benzoic acid
Formel: C₉H₈ClFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20483784
CAS-Nummer: 334652-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR