2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC32922-1G
PC32922-250MG
PC32922-5G
APOSPC32922-1GEA
58
EUR
APOSPC32922-1G
APOSPC32922-250MG
APOSPC32922-5G
2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole
Formel:
C₈H₄BrClF₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922505 CAS-Nummer: 1353778-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...