2-(Trifluoromethyl)-1H-benzimidazol-4-amine

Lieferant: Apollo Scientific
Gefahr

PC32933-250MG
APOSPC32933-250MGEA 364 EUR
APOSPC32933-250MG
2-(Trifluoromethyl)-1H-benzimidazol-4-amine
2-(Trifluoromethyl)-1H-benzimidazol-4-amine
Formel: C₈H₆F₃N₃
MDL: MFCD05856872
CAS-Nummer: 189045-18-9
UN: 2811

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR