6-Bromo-8-fluoro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC32965-100MG
APOSPC32965-100MGEA 31.9 EUR
APOSPC32965-100MG
6-Bromo-8-fluoro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
6-Bromo-8-fluoro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formel: C₉H₃BrF₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD32219447
CAS-Nummer: 2407339-59-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR