1-(3,4-Difluorophenyl)-imidazolidin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
PC32977-1G
PC32977-250MG
APOSPC32977-1GEA
84.1
EUR
APOSPC32977-1G
APOSPC32977-250MG
1-(3,4-Difluorophenyl)-imidazolidin-2-one
1-(3,4-Difluorophenyl)-imidazolidin-2-one
Formel:
C₉H₈F₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD23696668 CAS-Nummer: 162748-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...