1-(3,4-Difluorophenyl)-imidazolidin-2-one

Lieferant: Apollo Scientific
Achtung

PC32977-1G PC32977-250MG
APOSPC32977-1GEA 84.1 EUR
APOSPC32977-1G APOSPC32977-250MG
1-(3,4-Difluorophenyl)-imidazolidin-2-one
1-(3,4-Difluorophenyl)-imidazolidin-2-one
Formel: C₉H₈F₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD23696668
CAS-Nummer: 162748-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR