Perfluoro-compound FC-40™

Lieferant: Apollo Scientific
Achtung

Synonyme: FluorinertR(FC-40)

PC3312-1KG PC3312-250G
APOSPC3312-1KGEA 657 EUR
APOSPC3312-1KG APOSPC3312-250G
Perfluoro-compound FC-40™
Perfluoro-compound FC-40™

(Fluorinert is a registered trademark of 3M)

Formel: C₂₁F₄₈N₂
Molecular Weight: 1192.168 g/mol
Siedepunkt: 155 °C
Schmelzpunkt: –57 °C
Dichte: 1.87 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01632188
CAS-Nummer: 51142-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR