6-[3,5-Bis(trifluoromethyl)phenyl]-3-chloropyridazine

Lieferant: Apollo Scientific

PC3313-100MG PC3313-250MG
APOSPC3313-100MGEA 161 EUR
APOSPC3313-100MG APOSPC3313-250MG
6-[3,5-Bis(trifluoromethyl)phenyl]-3-chloropyridazine
6-[3,5-Bis(trifluoromethyl)phenyl]-3-chloropyridazine
Formel: C₁₂H₅ClF₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06801347
CAS-Nummer: 1031928-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR