3'-Fluoracetanilid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(3-Fluorphenyl)acetamid
PC3330-5G
APOSPC3330-5GEA
14.7
EUR
APOSPC3330-5G
3'-Fluoracetanilid 98%
3'-Fluoracetanilid
Formel:
C₈H₈FNO Molecular Weight: 153,16 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017917 CAS-Nummer: 351-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...