Perfluor-1-octen
Lieferant: Apollo Scientific
|
Achtung
|
PC3331-5G
APOSPC3331-5GEA
277
EUR
APOSPC3331-5G
Perfluor-1-octen
Perfluor-1-octen
Formel:
C₈F₁₆ Siedepunkt: 102…105 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00236708 CAS-Nummer: 559-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...