3,4-Difluor-5-(trifluormethyl)benzoesäure 95% min
Lieferant: Apollo Scientific
|
Achtung
|
PC3342-250MG
APOSPC3342-250MGEA
418
EUR
APOSPC3342-250MG
3,4-Difluor-5-(trifluormethyl)benzoesäure 95% min
3,4-Difluoro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃F₅O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236257 CAS-Nummer: 237761-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...