1H,1H-Perfluorohexyl-p-toluenesulphonate 97%

Lieferant: Apollo Scientific

PC3346-1G
APOSPC3346-1GEA 170 EUR
APOSPC3346-1G
1H,1H-Perfluorohexyl-p-toluenesulphonate 97%
1H,1H-Perfluorohexyl-p-toluenesulphonate
Formel: C₁₃H₉F₁₁O₃S
Molecular Weight: 454,26 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04038342
CAS-Nummer: 355-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR