Ethyl {[2-nitro-4-(trifluoromethyl)phenyl]amino}acetate
Lieferant: Apollo Scientific
PC3350-1G
APOSPC3350-1GEA
176
EUR
APOSPC3350-1G
Ethyl {[2-nitro-4-(trifluoromethyl)phenyl]amino}acetate
Ethyl {[2-nitro-4-(trifluoromethyl)phenyl]amino}acetate
Formel:
C₁₁H₁₁F₃N₂O₄ Molecular Weight: 292,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00239175 CAS-Nummer: 735-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...