2,6-Difluor-3-hydroxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

PC3351-250MG
APOSPC3351-250MGEA 0 EUR
APOSPC3351-250MG
2,6-Difluor-3-hydroxyphenylboronsäure
2,6-Difluor-3-hydroxyphenylboronsäure
Formel: C₆H₅BF₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD09878345
CAS-Nummer: 957065-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR