3-Brom-2-chlor-5-(trifluormethyl)pyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3352-25G
APOSPC3352-25GEA
88.6
EUR
APOSPC3352-25G
3-Brom-2-chlor-5-(trifluormethyl)pyridin 98%
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₂BrClF₃N Schmelzpunkt: 28…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878432 CAS-Nummer: 71701-92-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...