2,6-Difluor-4-hydroxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC3353-250MG
APOSPC3353-250MGEA
0
EUR
APOSPC3353-250MG
2,6-Difluor-4-hydroxyphenylboronsäure
2,6-Difluor-4-hydroxyphenylboronsäure
Formel:
C₆H₅BF₂O₃ Schmelzpunkt: 157 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09878344 CAS-Nummer: 957065-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...