4-Brom-N,N-dimethyl-2-fluorbenzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-fluoro-N,N-dimethylbenzamide
PC3354-1G
PC3354-5G
APOSPC3354-1GEA
94.4
EUR
APOSPC3354-1G
APOSPC3354-5G
4-Brom-N,N-dimethyl-2-fluorbenzamid 98%
4-Brom-N,N-dimethyl-2-fluorbenzamid
Formel:
C₉H₉BrFNO Molecular Weight: 246.0763 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878360 CAS-Nummer: 749927-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...