4-Brom-N,N-dimethyl-2-fluorbenzamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-fluoro-N,N-dimethylbenzamide

PC3354-1G PC3354-5G
APOSPC3354-1GEA 94.4 EUR
APOSPC3354-1G APOSPC3354-5G
4-Brom-N,N-dimethyl-2-fluorbenzamid 98%
4-Brom-N,N-dimethyl-2-fluorbenzamid
Formel: C₉H₉BrFNO
Molecular Weight: 246.0763 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878360
CAS-Nummer: 749927-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR