Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat

Lieferant: Apollo Scientific

PC3358-100MG
APOSPC3358-100MGEA 306 EUR
APOSPC3358-100MG
Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat
Ethyl-6,7-dichlor-3-(trifluormethyl)-1,4-dihydrochinoxalin-2-carboxylat
Formel: C₁₂H₉Cl₂F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09878367
CAS-Nummer: 957062-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR