3,3-Dimethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]azetidin-2-one

Lieferant: Apollo Scientific
Achtung

PC3362-250MG
APOSPC3362-250MGEA 340 EUR
APOSPC3362-250MG
3,3-Dimethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]azetidin-2-one
3,3-Dimethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]azetidin-2-one
Formel: C₁₂H₁₁F₃N₂O₃
Molecular Weight: 288,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907677
CAS-Nummer: 861881-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR