2,2,3,3-Tetrafluoropropyl trifluoromethanesulphonate 97%

Lieferant: Apollo Scientific
Gefahr

PC3383-50G
APOSPC3383-50GEA 1160 EUR
APOSPC3383-50G
2,2,3,3-Tetrafluoropropyl trifluoromethanesulphonate 97%
2,2,3,3-Tetrafluoropropyl trifluoromethanesulphonate
Formel: C₄H₃F₇O₃S
Siedepunkt: 122…125 °C/737mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02093338
CAS-Nummer: 6401-02-1
UN: 2927
ADR: 6.1+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR