2-(Trifluoroacetyl)pyrrole 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC3389-25G
PC3389-5G
PC3389-1G
APOSPC3389-25GEA
548
EUR
APOSPC3389-25G
APOSPC3389-5G
APOSPC3389-1G
2-(Trifluoroacetyl)pyrrole 99%
2-(Trifluoroacetyl)pyrrole
Formel:
C₆H₄F₃NO Molecular Weight: 163.100 g/mol Schmelzpunkt: 48…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141809 CAS-Nummer: 2557-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...